Skip to Content
MilliporeSigma
All Photos(1)

Documents

P2672

Sigma-Aldrich

Poly(ethylene glycol) diglycidyl ether

~75%

Synonym(s):

Diepoxy PEG, PEG diglycidyl ether, Polyoxyethylene bis(glycidyl ether)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C3H5O2-(C2H4O)n-C3H5O
CAS Number:
MDL number:
UNSPSC Code:
12161900

description

non-ionic

Assay

~75%

density

1.14 g/mL at 25 °C (lit.)

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Prepared from PEG with an average molecular weight of 3,350

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service