Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P2530

Sigma-Aldrich

[Tyr43]-PTH Fragment 43-68 human

≥97% (HPLC)

Synonym(s):

Tyr-Arg-Asp-Ala-Gly-Ser-Gln-Arg-Pro-Arg-Lys-Lys-Glu-Asp-Asn-Val-Leu-Val-Glu-Ser-His-Glu-Lys-Ser-Leu-Gly

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C126H208N42O43
CAS Number:
Molecular Weight:
2999.26
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

UniProt accession no.

storage temp.

−20°C

SMILES string

CC(C)CC(NC(=O)C(CO)NC(=O)C(CCCCN)NC(=O)C(CCC(O)=O)NC(=O)C(Cc1cnc[nH]1)NC(=O)C(CO)NC(=O)C(CCC(O)=O)NC(=O)C(NC(=O)C(CC(C)C)NC(=O)C(NC(=O)C(CC(N)=O)NC(=O)C(CC(O)=O)NC(=O)C(CCC(O)=O)NC(=O)C(CCCCN)NC(=O)C(CCCCN)NC(=O)C(CCCNC(N)=N)NC(=O)C2CCCN2C(=O)C(CCCNC(N)=N)NC(=O)C(CCC(N)=O)NC(=O)C(CO)NC(=O)CNC(=O)C(C)NC(=O)C(CC(O)=O)NC(=O)C(CCCNC(N)=N)NC(=O)C(N)Cc3ccc(O)cc3)C(C)C)C(C)C)C(=O)NCC(O)=O

Gene Information

human ... PTH(5741)

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Orwoll et al.
The Journal of laboratory and clinical medicine, 105(2), 247-253 (1985-02-01)
Parathyroid hormone (PTH) radioimmunoassays have conventionally utilized [125I]bPTH1-84 as a radioligand, but more region-specific PTH assays are now possible with the use of recently available synthetic PTH peptides as standards and radioligands. A radioiodination procedure has been developed that utilizes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service