Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P2419

Sigma-Aldrich

Monoclonal Anti-Peroxidase antibody produced in mouse

clone P6-38, ascites fluid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

P6-38, monoclonal

contains

15 mM sodium azide

species reactivity

horseradish

technique(s)

indirect ELISA: 1:5,000 using mouse primary antibody, bridging antibody and horseradish peroxidase

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Monoclonal anti-peroxidase (mouse IgG1 isotype) is derived from the hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse.

Immunogen

horseradish peroxidase

Application

Monoclonal Anti-Peroxidase antibody may be used for:
  • immunocytochemistry
  • indirect ELISA (1:5,000)
  • immunohistochemistry
  • immunoblotting

Biochem/physiol Actions

Peroxidase is often used as an enzyme label for antibodies during analyses of target antigens. Peroxidases aids in substrate conversion indicated by the formation of a coloured product. In an alternative procedure, an anti-enzyme antibody can be used to bridge a primary antibody and enzyme complex for the study of target proteins. Anti-peroxidase antibodies are also used to prepare peroxidase-anti-peroxidase (PAP) soluble complexes. Monoclonal Anti-Peroxidase antibody can be used for signal amplification in a wide range of immunological techniques. The product is specific for horseradish peroxidase. Such immunoassays are useful tools to detect antigens, toxins, bacteria and viruses.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leonid P Nezlin et al.
The Journal of comparative neurology, 464(3), 257-268 (2003-08-06)
Structural and functional investigations were carried out to study olfactory glomeruli in the main olfactory bulb (OB) in tadpoles of the clawed frog, Xenopus laevis. Calcium imaging of odor response patterns of OB neurons revealed that the synapses within the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service