Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P2238

Sigma-Aldrich

PBDA

>95% (NMR), liquid

Synonym(s):

4,4′-Phosphinicobis(butane-1,3-dicarboxylic acid)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H19O10P
CAS Number:
Molecular Weight:
354.25
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

>95% (NMR)

form

liquid

solubility

water: soluble

storage temp.

−20°C

SMILES string

OC(=O)CCC(CP(O)(=O)CC(CCC(O)=O)C(O)=O)C(O)=O

InChI

1S/C12H19O10P/c13-9(14)3-1-7(11(17)18)5-23(21,22)6-8(12(19)20)2-4-10(15)16/h7-8H,1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20)(H,21,22)

InChI key

SGRJLTOOQLQDRH-UHFFFAOYSA-N

Application

Potent inhibitor of glutamate carboxypeptidase II (NAAG peptidase), selective mGluR3-metabotropic glutamate receptor- agonist.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dual function glutamate-related ligands: discovery of a novel, potent inhibitor of glutamate carboxypeptidase II possessing mGluR3 agonist activity.
F Nan et al.
Journal of medicinal chemistry, 43(5), 772-774 (2000-03-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service