Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P1883

Sigma-Aldrich

L-Phenylalaninamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12N2O
CAS Number:
Molecular Weight:
164.20
Beilstein:
6270941
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

Assay

≥98%

form

powder

color

white to off-white

mp

92.8 °C

application(s)

detection

SMILES string

N[C@@H](Cc1ccccc1)C(N)=O

InChI

1S/C9H12N2O/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6,10H2,(H2,11,12)/t8-/m0/s1

InChI key

OBSIQMZKFXFYLV-QMMMGPOBSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

L-Phenylalaninamide is a chiral selector.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Mizanur Rahman et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 14(4), 1312-1321 (2007-11-23)
The monomer N'-octadecyl-N(alpha)-(4-vinyl)-benzoyl-L-phenylalanineamide (4) based on L-phenylalanine has been simply but effectively synthesized, and its self-assembling properties have been investigated. FTIR and a variable-temperature (1)H NMR spectroscopic investigation demonstrated that the aggregation of compound 4 in various organic solvents is
J F Rehfeld
Clinical chemistry, 44(5), 991-1001 (1998-05-20)
Shortage of reliable plasma assays has hampered studies of cholecystokinin (CCK). The assay problems are low plasma concentrations, extensive molecular heterogeneity, and close homology of CCK to gastrin, which circulates in higher concentrations. To develop an accurate CCK RIA, antibodies
Lenka Maletínská et al.
Peptides, 32(9), 1887-1892 (2011-08-30)
Prolactin-releasing peptide (PrRP)-induced secretion of prolactin is not currently considered a primary function of PrRP, but the development of late-onset obesity in both PrRP and PrRP receptor knock-out mice indicates the unique anorexigenic properties of PrRP. In our recent study
K Morihara
Biomedica biochimica acta, 50(10-11), S15-S18 (1991-01-01)
The semisynthesis of C-terminal peptides of gastrin, calcitonin gene-related peptides (rat and human), and cholecystokinin, or of human neuropeptide Y was achieved by introduction of Phe-NH2 or Tyr-NH2 at their carboxyl termini, respectively. Thermolysin or the related enzyme was used
Seon-Yeong Kwak et al.
Bioorganic & medicinal chemistry letters, 20(2), 738-741 (2009-12-08)
Kojic acid-phenylalanine amide (KA-F-NH(2)), which showed an excellent tyrosinase inhibitory activity, did not inhibit melanogenesis in melanocyte due to its low cell permeability. To enhance its cell permeability by increasing lipophilicity, we prepared metal coordination compounds of KA-F-NH(2) and characterized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service