Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P1849

Supelco

Pinacidil monohydrate

Synonym(s):

(±)-N-Cyano-N′-4-pyridinyl-N″-(1,2,2-trimethylpropyl)guanidine monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H19N5 · H2O
CAS Number:
Molecular Weight:
263.34
MDL number:
UNSPSC Code:
41116107
Pricing and availability is not currently available.

SMILES string

O.CC(N\C(Nc1ccncc1)=N\C#N)C(C)(C)C

InChI

1S/C13H19N5.H2O/c1-10(13(2,3)4)17-12(16-9-14)18-11-5-7-15-8-6-11;/h5-8,10H,1-4H3,(H2,15,16,17,18);1H2

InChI key

AFJCNBBHEVLGCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Potassium channel activator; antihypertensive

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service