Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P1655

Sigma-Aldrich

Purine

Synonym(s):

7H-Imidazo(4,5-d)pyrimidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4N4
CAS Number:
Molecular Weight:
120.11
Beilstein:
3200
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥98%

mp

214-217 °C (lit.)

storage temp.

−20°C

SMILES string

c1ncc2nc[nH]c2n1

InChI

1S/C5H4N4/c1-4-5(8-2-6-1)9-3-7-4/h1-3H,(H,6,7,8,9)

InChI key

KDCGOANMDULRCW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Packaged under Argon

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benshang Li et al.
Nature medicine, 21(6), 563-571 (2015-05-12)
Relapse is the leading cause of mortality in children with acute lymphoblastic leukemia (ALL). Among chemotherapeutics, thiopurines are key drugs in ALL combination therapy. Using whole-exome sequencing, we identified relapse-specific mutations in the phosphoribosyl pyrophosphate synthetase 1 gene (PRPS1), which
Silvia Meneghesso et al.
ChemMedChem, 8(3), 415-425 (2013-02-07)
2'-Fluoro-2'-deoxyguanosine has been reported to have potent anti-influenza virus activity in vitro and in vivo. Herein we describe the synthesis and biological evaluation of 6-modified 2'-fluoro-2'-deoxyguanosine analogues and their corresponding phosphoramidate ProTides as potential anti-influenza virus agents. Whereas the parent
Sarah C Hennebry et al.
PloS one, 7(12), e46675-e46675 (2013-01-04)
The transthyretin-like protein (TLP) from Salmonella enterica subspecies I is a periplasmic protein with high level structural similarity to a protein found in mammals and fish. In humans, the protein homologue, transthyretin, binds and carries retinol and thyroxine, and a
Elijah Oyoo-Okoth et al.
Aquatic toxicology (Amsterdam, Netherlands), 124-125, 34-40 (2012-08-14)
Infestation of fish by endoparasites may potentially influence metal uptake and elimination by the host. We quantified the metal uptake rate constant (k(u)) and efflux rate constants (k(e)) of radiolabeled Cd and Co in the cyprinid fish Rastrineobola argentea experimentally
Hiroshi Ashihara et al.
Phytochemistry, 69(4), 841-856 (2007-12-11)
Details of the recently elucidated biosynthetic pathways of caffeine and related purine alkaloids are reviewed. The main caffeine biosynthetic pathway is a sequence consisting of xanthosine-->7-methylxanthosine-->7-methylxanthine-->theobromine-->caffeine. Genes encoding N-methyltransferases involved in three of these four reactions have been isolated and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service