Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

P154

Sigma-Aldrich

Pinacidil monohydrate

powder

Synonym(s):

(±)-N-Cyano-N′-4-pyridinyl-N″-(1,2,2-trimethylpropyl)guanidine monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H19N5 · H2O
CAS Number:
Molecular Weight:
263.34
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

form

powder

Quality Level

color

white to beige

solubility

DMSO: 15 mg/mL, clear
45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 40 mg/mL

SMILES string

O.CC(N\C(Nc1ccncc1)=N\C#N)C(C)(C)C

InChI

1S/C13H19N5.H2O/c1-10(13(2,3)4)17-12(16-9-14)18-11-5-7-15-8-6-11;/h5-8,10H,1-4H3,(H2,15,16,17,18);1H2

InChI key

AFJCNBBHEVLGCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Pinacidil monohydrate has been used:
  • in repolarization studies by infusing into left anterior descending artery (LAD)
  • to induce ATP-sensitive K+ (KATP) current in ventricular myocytes for electrophysiological studies
  • as a KATP opener to treat myocardial cells

Biochem/physiol Actions

K+ channel activator; antihypertensive.
Pinacidil is a KATP channel agonist. It protects cardiomyocytes by preventing loss of mitochondrial membrane potential. It prevents the caspase 3 activation in hypoxia/reoxygenation injury.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun-Yih Chen et al.
Shock (Augusta, Ga.), 38(2), 203-212 (2012-05-12)
Alterations in the activity of vascular K channels are commonly associated with abnormalities in cerebral vascular function after subarachnoid hemorrhage (SAH). Subarachnoid hemorrhage-induced vasospasm remains incompletely understood; nevertheless, activation of K channels may be of benefit in relieving spastic constriction.
Milla Mikkola et al.
Stem cells and development, 22(5), 707-716 (2012-10-31)
Lectins are carbohydrate-binding proteins, which occur ubiquitously in nature and are abundant in all living organisms from bacteria to mammals. They have several biological functions among which cell adhesion is well known and characterized. Based on the characterization of the
Asami Mori et al.
European journal of pharmacology, 669(1-3), 94-99 (2011-08-30)
The large-conductance Ca(2+)-activated K(+) (BK(Ca)) channels modulate the retinal vascular tone, but question of whether the impairment of the channel function contributes to abnormalities of retinal circulation has not yet been completely elucidated. The purpose of this study was to
Protective actions of epoxyeicosatrienoic acid: dual targeting of cardiovascular PI3K and KATP channels
Bodiga S, et al.
Journal of Molecular and Cellular Cardiology, 46(6), 978-988 (2009)
Ivana Barbaric et al.
Cryobiology, 63(3), 298-305 (2011-10-27)
Human embryonic stem cells (hESCs) can be maintained as undifferentiated cells in vitro and induced to differentiate into a variety of somatic cell types. Thus, hESCs provide a source of differentiated cell types that could be used to replace diseased

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service