Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

P1510

Sigma-Aldrich

Petroselinyl alcohol

liquid

Synonym(s):

cis-6-Octadecen-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H36O
CAS Number:
Molecular Weight:
268.48
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.25

form

liquid

storage temp.

−20°C

SMILES string

CCCCCCCCCCC\C=C/CCCCCO

InChI

1S/C18H36O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h12-13,19H,2-11,14-18H2,1H3/b13-12-

InChI key

TVPWKOCQOFBNML-SEYXRHQNSA-N

Packaging

Sealed ampule.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service