Skip to Content
MilliporeSigma

P1402

1-O-Palmityl-2-acetyl-rac-glycero-3-phosphocholine

~99%

Synonym(s):

rac-Platelet activating factor, DL-α-Phosphatidylcholine, β-acetyl-γ-O-hexadecyl

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C26H54NO7P
CAS Number:
Molecular Weight:
523.68
UNSPSC Code:
12352211
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

~99%

form

solid

functional group

phospholipid

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCOCC(COP(O)(=O)OCC[N](C)(C)C)OC(C)=O

InChI

1S/C26H55NO7P/c1-6-7-8-9-10-11-12-13-14-15-16-17-18-19-21-31-23-26(34-25(2)28)24-33-35(29,30)32-22-20-27(3,4)5/h26H,6-24H2,1-5H3,(H,29,30)

InChI key

LMEXBIODJFQAQA-UHFFFAOYSA-N



Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library