Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P1294

Sigma-Aldrich

1,10-Phenanthroline monohydrate

ACS reagent, passes test for redox indicator, passes test for iron determination

Synonym(s):

o-Phenanthroline monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8N2 · H2O
CAS Number:
Molecular Weight:
198.22
Beilstein:
4008096
EC Number:
MDL number:
UNSPSC Code:
12352100

grade

ACS reagent

mp

100-104 °C (lit.)

suitability

passes test for iron determination
passes test for redox indicator

SMILES string

O.c1cnc2c(c1)ccc3cccnc23

InChI

1S/C12H8N2.H2O/c1-3-9-5-6-10-4-2-8-14-12(10)11(9)13-7-1;/h1-8H;1H2

InChI key

PPQJCISYYXZCAE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

When complexed with copper, it possesses nuclease activity that has been used to study DNA-protein interactions.

Biochem/physiol Actions

Metalloprotease inhibitor; chelates iron, zinc and other divalent metals. Effective concentration 1-10 mM.

replaced by

Product No.
Description
Pricing

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ayman A Gouda et al.
Chemistry Central journal, 2, 7-7 (2008-04-16)
Etodolac (ETD) is a non-steroidal anti-inflamatory antirheumatic drug. A survey of the literature reveals that there is no method available for the determination of ETD in pure form and pharmaceutical formulations by oxidation-reduction reactions. We describe three simple, sensitive and
Linlin Yang et al.
Analytical biochemistry, 492, 56-62 (2015-09-26)
We demonstrate a novel protocol for sensitive in situ label-free electrochemical detection of DNA hybridization based on copper complex ([Cu(phen)2](2+), where phen = 1,10-phenanthroline) and graphene (GR) modified glassy carbon electrode. Here, [Cu(phen)2](2+) acted advantageously as both the electrochemical indicator and the
Hongwei Chen et al.
ACS applied materials & interfaces, 7(23), 12814-12823 (2015-05-27)
Although many techniques exist for fabricating near-infrared (NIR)-resonant and magnetic resonance imaging (MRI)-capable nanomediators for photothermal cancer therapy, preparing them in an efficient and scalable process remains a significant challenge. In this report, we exploit one-step siloxane chemistry to facilely
Sarah Louise Dombernowsky et al.
Nature communications, 6, 7518-7518 (2015-06-26)
The metalloproteinase ADAM17 activates ErbB signalling by releasing ligands from the cell surface, a key step underlying epithelial development, growth and tumour progression. However, mechanisms acutely controlling ADAM17 cell-surface availability to modulate the extent of ErbB ligand release are poorly
Björn Harink et al.
Electrophoresis, 36(3), 475-484 (2014-09-30)
We present here a screening method based on a microfluidic platform, which can generate four orthogonal and overlapping concentration gradients of soluble compounds over a monolayer of cells, in combination with automated and in situ image analysis, for use in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service