P119
(±)-Pindobind
solid
Synonym(s):
N8-Bromoacetyl-N1-3′-(4-indolyloxy)-2′-hydroxy-propyl-(Z)-1,8-diamino-p-menthane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C23H34BrN3O3
CAS Number:
Molecular Weight:
480.44
MDL number:
UNSPSC Code:
12352103
form
solid
color
white
solubility
DMSO: soluble
ethanol: soluble
methanol: soluble
storage temp.
−20°C
Biochem/physiol Actions
Analog of pindolol that contains a bromo-acetyl group capable of alkylating β-adrenergic receptors; easily radioiodinated.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
R J Box
Journal of receptor research, 9(4-5), 385-403 (1989-01-01)
The reactivities with beta-adrenergic receptors of the bromoacetyl derivatives of the beta-adrenergic antagonists alprenolol and pindolol, BAAM and BIM, respectively, were compared in intact S49 mouse lymphoma cells. Both compounds caused irreversible blockade of receptors and changes in the mobility
J Pitha et al.
Journal of medicinal chemistry, 30(4), 612-615 (1987-04-01)
New alkylating ligands derived from indole with high affinity for beta-adrenoceptors were synthesized and their properties examined. N8-(Bromoacetyl)-N1-[3-(4-indolyloxy)-2-hydroxypropyl]-(Z)-1,8-dia mino-p- menthane (8) and its N1,N8 isomer (9) were prepared by the reaction of bromoacetyl bromide with a product of the condensation
P Molenaar et al.
Biochemical pharmacology, 37(19), 3601-3607 (1988-10-01)
The actions of alkylating pindolol (N8-bromoacetyl-N1-3'-(4-indolyloxy)-2'-hydroxypropyl[z]-1,8- diamino-p-menthane; BIM) have been examined on beta-adrenoceptors in guinea-pig left atria and trachea. In organ bath experiments, addition of BIM (greater than or equal to 0.1 microM), followed by washout, produced concentration-dependent rightward shifts
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service