Skip to Content
MilliporeSigma
All Photos(1)

Documents

P1131

Supelco

Citric acid sesquipiperazine salt hydrate

Synonym(s):

tri-Piperazine dicitrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C4H10N2)3 · (C6H8O7)2 · xH2O
CAS Number:
Molecular Weight:
321.33 (anhydrous basis)
Beilstein:
3777748
EC Number:
MDL number:
UNSPSC Code:
41116107

SMILES string

OC(C(CC(O)=O)(CC(O)=O)O)=O.OC(C(CC(O)=O)(O)CC(O)=O)=O.O.N1CCNCC1.N2CCNCC2.N3CCNCC3

InChI

1S/2C6H8O7.3C4H10N2.H2O/c2*7-3(8)1-6(13,5(11)12)2-4(9)10;3*1-2-6-4-3-5-1;/h2*13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);3*5-6H,1-4H2;1H2

InChI key

LWMBPKJYEQGDLN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

comparable product

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service