Skip to Content
MilliporeSigma
All Photos(1)

Documents

P107

Sigma-Aldrich

N6-(2-Phenylethyl)adenosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21N5O4
CAS Number:
Molecular Weight:
371.39
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

form

solid

solubility

0.1 M NaOH: soluble
H2O: very slightly soluble
ethanol: moderately soluble

SMILES string

OCC1OC(C(O)C1O)n2cnc3c(NCCc4ccccc4)ncnc23

InChI

1S/C18H21N5O4/c24-8-12-14(25)15(26)18(27-12)23-10-22-13-16(20-9-21-17(13)23)19-7-6-11-4-2-1-3-5-11/h1-5,9-10,12,14-15,18,24-26H,6-8H2,(H,19,20,21)

InChI key

LGZYEDZSPHLISU-UHFFFAOYSA-N

Gene Information

human ... ADORA1(134)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Selective A1 adenosine receptor agonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Debora Bencivenga et al.
Molecular oncology, 15(4), 915-941 (2020-12-15)
CDKN1B haploinsufficiency promotes the development of several human cancers. The gene encodes p27Kip1 , a protein playing pivotal roles in the control of growth, differentiation, cytoskeleton dynamics, and cytokinesis. CDKN1B haploinsufficiency has been associated with chromosomal or gene aberrations. However

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service