P1003
O-Phospho-DL-threonine
Synonym(s):
DL-2-Amino-3-hydroxybutanoic acid 3-phosphate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C4H10NO6P
CAS Number:
Molecular Weight:
199.10
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
Recommended Products
Assay
≥97.0% (TLC)
form
powder
color
white
storage temp.
−20°C
SMILES string
CC(OP(O)(O)=O)C(N)C(O)=O
InChI
1S/C4H10NO6P/c1-2(3(5)4(6)7)11-12(8,9)10/h2-3H,5H2,1H3,(H,6,7)(H2,8,9,10)
InChI key
USRGIUJOYOXOQJ-UHFFFAOYSA-N
Biochem/physiol Actions
O-Phospho-DL-threonine is an amino acid derivative.
Signal Word
Warning
Hazard Statements
Hazard Classifications
Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
LipA, a Tyrosine and Lipid Phosphatase Involved in the Virulence of Listeria monocytogenes
Kastner, R. et al.
Infection and Immunity, 29, 2489-2498 (2011)
Jean-Yves Winum et al.
Bioorganic & medicinal chemistry letters, 15(6), 1683-1686 (2005-03-05)
The interaction of five human carbonic anhydrase (hCA, EC 4.2.1.1) isozymes, that is, hCA I, II, IV, V, and IX with a small library of phosphonic acids/organic phosphates, including methylphosphonic acid, MPA; phenylphosphonic acid, PPA; N-(phosphonoacetyl)-L-aspartic acid, PALA, methylene diphosphonic
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service