Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P0937

Sigma-Aldrich

Poly (α,β-[N-(3-hydroxypropyl)-DL-aspartamide])

≥95% (TLC), Mol wt 5,000-20,000

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.26

Product Name

Poly (α,β-[N-(3-hydroxypropyl)-DL-aspartamide]), ≥95% (TLC), mol wt 5,000-20,000

Assay

≥95% (TLC)

form

solid

mol wt

5,000-20,000

storage temp.

−20°C

Biochem/physiol Actions

Poly (α,β-[N-(3-hydroxypropyl)-DL-aspartamide]) as a drug carrier has shown sustained drug release for up to 120 days.

Preparation Note

Prepared via polysuccinimide.

Analysis Note

Molecular weight based on viscosity.

Other Notes

For additional technical information on polyamino acids please visit the Polyamino acid FAQ resource.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Poly (α,β-[N-(3-hydroxypropyl)-DL-aspartamideA:a new drug carrier
Tang, Q., Zhu, K.J., Chen, Q. Q.
Journal of Applied Polymer Science. Applied Polymer Symposium, 77, 2411-2417 (2000)
Synthesis of alpha beta-poly((2-hydroxyethyl)-DL-aspartamide), a new plasma expander.
P Neri et al.
Journal of medicinal chemistry, 16(8), 893-897 (1973-08-01)

Articles

Humankind has utilized protein materials throughout its existence, starting with the use of materials such as wool and silk for warmth and protection from the elements and continuing with the use of recombinant DNA techniques to synthesize proteins with unique and useful properties.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service