Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P0740

Sigma-Aldrich

N-(2-Phenylethyl)indomethacinamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H25N2O3Cl
CAS Number:
Molecular Weight:
460.95
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (TLC)

Quality Level

form

solid

mp

158-162 °C

shipped in

wet ice

storage temp.

−20°C

SMILES string

COc1ccc2n(c(C)c(CC(=O)NCCc3ccccc3)c2c1)C(=O)c4ccc(Cl)cc4

InChI

1S/C27H25ClN2O3/c1-18-23(17-26(31)29-15-14-19-6-4-3-5-7-19)24-16-22(33-2)12-13-25(24)30(18)27(32)20-8-10-21(28)11-9-20/h3-13,16H,14-15,17H2,1-2H3,(H,29,31)

InChI key

VYDBTNADENXYSN-UHFFFAOYSA-N

Biochem/physiol Actions

Inhibits human recombinant and ovine cyclooxygenase-2 (COX-2) with IC50s of 0.06 μM and 0.125 μM, respectively; over 400-fold less potent as an inhibitor of COX-1.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service