P0514
L-α-Phosphatidyl-DL-glycerol ammonium salt from egg yolk lecithin
solution, 10 mg/mL in chloroform: methanol (49:1), ≥99%
Synonym(s):
1,2-Diacyl-sn-glycero-3-phospho-(1-rac-glycerol), 1-(3-sn-Phosphatidyl)-rac-glycerol ammonium salt from egg yolk lecithin
About This Item
Recommended Products
biological source
egg yolk
Quality Level
Assay
≥99%
form
solution
concentration
10 mg/mL in chloroform: methanol (49:1)
functional group
phospholipid
lipid type
phosphoglycerides
storage temp.
−20°C
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3
Target Organs
Central nervous system, Liver,Kidney
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service