Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P0415

Sigma-Aldrich

Potassium cyanide-14C

solid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K14CN
CAS Number:
Molecular Weight:
65.12
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

extent of labeling

40-60 mCi per mmol

mass shift

M+2

storage temp.

2-8°C

SMILES string

[K][14CH2]C#N

InChI

1S/C2H2N.K/c1-2-3;/h1H2;/i1+2;

InChI key

IJAJWWZKTPVZIH-DEQYMQKBSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Sealed ampule

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Oral - Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Met. Corr. 1 - STOT RE 1

Target Organs

Thyroid

Supplementary Hazards

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service