Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P0175

Sigma-Aldrich

pGlu-Val-Asn-Phe-Ser-Pro-Gly-Trp-Gly-Thr-NH2

≥95% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C50H67N13O14
CAS Number:
Molecular Weight:
1074.15
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.26

product name

pGlu-Val-Asn-Phe-Ser-Pro-Gly-Trp-Gly-Thr-NH2, ≥95% (HPLC)

Assay

≥95% (HPLC)

form

powder

color

white to off-white

storage temp.

−20°C

SMILES string

CC(C)C(NC(=O)C1CCC(=O)N1)C(=O)NC(CC(N)=O)C(=O)NC(Cc2ccccc2)C(=O)NC(CO)C(=O)N3CCCC3C(=O)NCC(=O)NC(Cc4c[nH]c5ccccc45)C(=O)NCC(=O)NC(C(C)O)C(N)=O

InChI

1S/C50H67N13O14/c1-25(2)41(62-45(72)31-15-16-38(67)56-31)49(76)59-34(20-37(51)66)47(74)58-32(18-27-10-5-4-6-11-27)46(73)60-35(24-64)50(77)63-17-9-14-36(63)48(75)55-22-39(68)57-33(19-28-21-53-30-13-8-7-12-29(28)30)44(71)54-23-40(69)61-42(26(3)65)43(52)70/h4-8,10-13,21,25-26,31-36,41-42,53,64-65H,9,14-20,22-24H2,1-3H3,(H2,51,66)(H2,52,70)(H,54,71)(H,55,75)(H,56,67)(H,57,68)(H,58,74)(H,59,76)(H,60,73)(H,61,69)(H,62,72)

InChI key

NHXHBXCOYZFUSO-UHFFFAOYSA-N

Amino Acid Sequence

Glp-Val-Asn-Phe-Ser-Pro-Gly-Trp-Gly-Thr-NH2

Biochem/physiol Actions

Hypertrehalosemic neuropeptide from cockroach (Nauphoeta cinerea) with an amino acid sequence similar to that of adipokinetic hormone; involved in carbohydrate metabolism and increases the trehalose content of hemolymph.
Glp-Val-Asn-Phe-Ser-Pro-Gly-Trp-Gly-Thr-NH2 is a hypertrehalosemic neuropeptide (Nauphoeta cinerea) has an amino acid sequence similar to that of adipokinetic hormone and is involved in carbohydrate metabolism and increases the trehalose content of hemolymph.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Gäde et al.
General and comparative endocrinology, 86(1), 119-127 (1992-04-01)
Two hypertrehalosemic neuropeptides from the corpus cardiacum of the cockroach Polyphaga aegyptiaca were isolated by reversed-phase high-performance liquid chromatography, and their primary structures were determined by pulsed-liquid phase sequencing employing Edman chemistry, after enzymically deblocking the N-terminal pyroglutamate residue. As
J Y Bradfield et al.
Proceedings of the National Academy of Sciences of the United States of America, 88(10), 4558-4562 (1991-05-15)
Hypertrehalosemic hormone (a carbohydrate-mobilizing neuroendocrine decapeptide) and starvation markedly increased levels of a cockroach (Blaberus discoidalis) fat body cytochrome P450 message. The gene represented by the cloned P450 cDNA has been named CYP4C1 (cytochrome P450 family 4, subfamily C, gene
G Gäde et al.
Biochemical and biophysical research communications, 141(2), 774-781 (1986-12-15)
The hypertrehalosaemic factor from the cockroach Nauphoeta cinerea, a decapeptide, has been assigned the structure >Glu-Val-Asn-Phe-Ser-Pro-Gly-Trp-Gly-Thr-NH2. The structure was assigned from its high-resolution fast atom bombardment mass spectrum and metastable studies on the M + H ion and confirmed by
J Kim et al.
Biochemistry and molecular biology international, 35(4), 803-811 (1995-04-01)
A new method for the determination of the modified amino acid structures of peptides containing pyroglutamyl residue in their N-terminal position is reported. The combination of fast atom bombardment (FAB) mass spectrometry and pyroglutamate aminopeptidase digestion provides a convenient and
Role of Insect Neuropeptides and Juvenile Hormones in Silk Protein Biosynthesis
Unni, B.G. et al.
Annals of Neurology, 15, 112-117 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service