Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P0083

Sigma-Aldrich

PRMT1 from rat

recombinant, expressed in E. coli, ≥90% (SDS-PAGE), buffered aqueous solution

Synonym(s):

HMT1-like 2, HRMT1L2, Heterogenious nuclear ribonucleoprotein methyltransferase 1-like 2, IR1B4, Interferon receptor 1-bound protein 4, Protein arginine N-methyl transferase 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51111800
NACRES:
NA.32

recombinant

expressed in E. coli

Quality Level

Assay

≥90% (SDS-PAGE)

form

buffered aqueous solution

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

Biochem/physiol Actions

Methyl transferases catalyze the addition of methyl groups to nitrogen, carbon, sulfur, and oxygen atoms of small molecules, lipids, proteins, and nucleic acids. Eight mammalian protein arginine methyltransferases (PRMT) have been identified. PRMT1 is the predominant member of the methyl transferases, which catalyzes the protein arginine N-methylation reactions. PRMT1 is implicated in various cellular processes including: transcription, RNA processing, and signal transduction.
PRMT1 is the predominant member of the methyl transferases, which catalyzes the protein arginine N-methylation reactions. PRMT1 is implicated in various cellular processes including: transcription, RNA processing, and signal transduction.

Unit Definition

The specific activity is ≥ 0.1 nmol/mgP/min. measured by 3H-AdoMet incorporation into histone for 30 minues at 30 °C.

Physical form

Solution of 50 mM Tris, pH 7.6, 5 mM DTT, 0.2% IGEPAL® CA-630, 150 mM NaCl, and 30% glycerol (w/v).

Analysis Note

The N-methyltransferase activity is determined by detecting the level of radiolabel transfer from 3H-AdoMet (Methyl donor) to histone (Cat. No. H4380), which is arginine rich (methyl acceptor).

Legal Information

IGEPAL is a registered trademark of Solvay

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hsin-Wei Liao et al.
The Journal of clinical investigation, 125(12), 4529-4543 (2015-11-17)
Posttranslational modifications to the intracellular domain of the EGFR are known to regulate EGFR functions; however, modifications to the extracellular domain and their effects remain relatively unexplored. Here, we determined that methylation at R198 and R200 of the EGFR extracellular
Xiaolan Deng et al.
Oncotarget, 6(34), 35173-35182 (2015-10-16)
Inner centromere protein (INCENP) is a part of a protein complex known as the chromosomal passenger complex (CPC) that is essential for correcting non-bipolar chromosome attachments and for cytokinesis. We here demonstrate that a protein arginine methyltransferase PRMT1, which are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service