Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P0057

Sigma-Aldrich

PPBP maleate

≥98% (HPLC), powder

Synonym(s):

4-PPBP maleate, 4-phenyl-1-(4-phenylbutyl)-piperidine maleate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27N · xC4H4O4
CAS Number:
Molecular Weight:
293.45 (free base basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

off-white

solubility

DMSO: >20 mg/mL

storage temp.

2-8°C

SMILES string

OC(=O)\C=C/C(O)=O.C(CCc1ccccc1)CN2CCC(CC2)c3ccccc3

InChI

1S/C21H27N.C4H4O4/c1-3-9-19(10-4-1)11-7-8-16-22-17-14-21(15-18-22)20-12-5-2-6-13-20;5-3(6)1-2-4(7)8/h1-6,9-10,12-13,21H,7-8,11,14-18H2;1-2H,(H,5,6)(H,7,8)/b;2-1-

InChI key

OASPNIMFGJVLES-BTJKTKAUSA-N

Biochem/physiol Actions

PPBP maleate is a potent σ1 receptor ligand.
PPBP maleate is a potent σ1 receptor ligand. PPBP is a very potent agonist of σ1 receptor (Ki = 0.8 nM).

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Sigma receptors may constitute favorable targets for drug design in conditions such as psychiatric and movement disorders, amnesia, depression, cancer, inflammation and cocaine addiction.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service