Skip to Content
MilliporeSigma
All Photos(1)

Documents

OH83033967

Ohaus® pH Electrode

ST320

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115604

material

epoxy shaft

packaging

pack of 1 ea

manufacturer/tradename

Ohaus® 83033967

pH measuring range

0.00-14 pH unit

temperature measuring range

0.0-80 °C

Looking for similar products? Visit Product Comparison Guide

General description

All electrodes include a temperature sensor, which powers automatic temperature compensation and ensures accurate measurements.

Application

pH, reference, oxidation-reduction potential (ORP), conductivity, dissolved oxygen (DO), temperature measurement

Features and Benefits

Can be used in conjunction with all Starter bench and portable meters

Legal Information

Ohaus is a registered trademark of Ohaus Corp.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service