Key Documents
OH30253059
Ohaus® Scout® Portable Touchscreen Balance
Model STX223, weighing capacity 220 g, resolution: 0.001 g, AC/DC input 230 V
About This Item
UNSPSC Code:
41111502
NACRES:
NB.13
material
metal weighing plate
feature
CE compliant
packaging
pack of 1 ea
manufacturer/tradename
Ohaus® 30253059
weighing capacity measuring range
220 g, resolution: 0.001 g
weighing pan diam.
9.3 cm
General description
Ideal for laboratory and industrial applications, the OHAUS Scout features a user-friendly color touchscreen and a compact stackable design. Designed to deliver superior performance with rapid stability testing and high resolution weighing results, these portable scales set a new standard in weighing applications. Their features also include superior overload protection and advanced connectivity options.
Application
Weighing, piece counting,% weighing, check weighing, animal weighing / dynamic weighing, totalizing, density determination, display hold, mole weighing
Features and Benefits
Stability indicator, stackable design with storage cover included (except 1 mg models), mechanical and software overload / underload protection, low battery level indicator, automatic shutdown, auto-tare, slope calibration mass included with models up to 620g.
Legal Information
Ohaus is a registered trademark of Ohaus Corp.
Scout is a registered trademark of Ohaus Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)