Skip to Content
MilliporeSigma

Skip To

OGS239

PSF-PA-PROMMCS-BETAGAL - NO PROMOTER (MCS) BETA GAL PLASMID

plasmid vector for molecular cloning

Synonym(s):

cloning vector, expression vector, molecular cloning vector, plasmid, plasmid vector, snapfast vector, vector

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.85
UNSPSC Code:
12352200
Origin of replication:
pUC (500 copies)
Bacteria selection:
kanamycin
Reporter gene:
none
Peptide cleavage:
no cleavage


form

buffered aqueous solution

mol wt

size 5395 bp

bacteria selection

kanamycin

origin of replication

pUC (500 copies)

peptide cleavage

no cleavage

reporter gene

none

shipped in

ambient

storage temp.

−20°C

General description

This plasmid contains a multiple cloning site in the promoter position designed to allow you to insert your own promoter to drive the expression of the reporter gene. In this plasmid it is upstream of the beta galactosidase (beta gal) reporter gene. The promoter multiple cloning site extends from the SalI restriction site to the BstBI restriction site. Downstream sites have other functions in our plasmid system for example adding n-terminal peptide tags.

Promoter Expression Level:

Application

Cloning in a gene: This plasmid has been designed to be compatible with a range of cloning techniques. The multiple cloning site contains a range of standard commonly used restriction sites for cloning. Using these sites genes can be inserted using standard cloning methods with DNA ligase. Other methods such as ligase independent cloning (LIC) Gibson Assembly InFusionHD or Seamless GeneArt can also be used and because all of our plasmids are based on the same backbone the same method can be used for cloning into all of our catalogue vectors.

Multiple cloning site notes: There are a few important sites within the MCS. These include the NcoI site the XbaI site and the BsgI and BseRI sites. The NcoI site contains a start codon that is immediately downstream of both a Kozak and Shine-Dalgarno ribosomal binding site. These allow for optimal positioning of genes when the start codon is placed in this location. If this is not required and you wish to use a downstream site for gene cloning you can remove the NcoI site by cleaving the plasmid with KpnI.

The XbaI site contains a stop codon. This stop codon is positioned in a specific position in relation to the BsgI and BseRI sites that are immediately downstream. When either BseRI or BsgI cleave the plasmid they produce a TA overhang from the stop codon in the XbaI site that is compatible with all of our peptide tag plasmids cut with the same sites. BseRI and BsgI sites are non-palindromic and cleave a defined number of bases away from their binding site.

Whenever we clone a gene into our multiple cloning site we always position the start and stop codon in the same positions in the MCS. If the start and ends of the genes are not compatible with NcoI and XbaI we extend the sequence to the nearest external sites but keep the start and stop codons locations consistent.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.oxgene.com

Other Notes

To view sequence information for this product, please visit the product page


Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters