Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

O9881

Sigma-Aldrich

Methyl cis-12-octadecenoate

~99% (capillary GC)

Synonym(s):

cis-12-Octadecenoic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H36O2
CAS Number:
Molecular Weight:
296.49
MDL number:
UNSPSC Code:
12352211

Assay

~99% (capillary GC)

storage temp.

−20°C

SMILES string

CCCCC\C=C/CCCCCCCCCCC(=O)OC

Looking for similar products? Visit Product Comparison Guide

Packaging

Sealed ampule, under Argon.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service