Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

O9875

Sigma-Aldrich

Oleyl acetate

≥97% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H38O2
CAS Number:
Molecular Weight:
310.51
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

≥97% (GC)

form

liquid

functional group

ester

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C/CCCCCCCCOC(C)=O

InChI

1S/C20H38O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-22-20(2)21/h10-11H,3-9,12-19H2,1-2H3/b11-10-

InChI key

GYGAZRPDUOHMAF-KHPPLWFESA-N

Packaging

Sealed ampule.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carmen Quero et al.
PloS one, 12(2), e0171166-e0171166 (2017-02-09)
Castniid moths (Lepidoptera: Castniidae) display a butterfly-like reproductive behavior, i.e., they use visual stimuli for mate location and females have apparently lost their pheromone glands in an evolutionary context. In this paper we report for the first time the identification

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service