Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

O5133

Sigma-Aldrich

Oligothymidylic acid d(pT)2 sodium salt

lyophilized powder

Synonym(s):

d(pT)2

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

Assay

≥90% (HPLC)

form

lyophilized powder

storage temp.

−20°C

SMILES string

[Na+].CC1=CN([C@H]2C[C@H](O)[C@@H](COP(O)(=O)O[C@H]3C[C@@H](O[C@@H]3COP(O)([O-])=O)N4C=C(C)C(=O)NC4=O)O2)C(=O)NC1=O

InChI

1S/C20H28N4O15P2.Na/c1-9-5-23(19(28)21-17(9)26)15-3-11(25)13(37-15)7-36-41(33,34)39-12-4-16(38-14(12)8-35-40(30,31)32)24-6-10(2)18(27)22-20(24)29;/h5-6,11-16,25H,3-4,7-8H2,1-2H3,(H,33,34)(H,21,26,28)(H,22,27,29)(H2,30,31,32);/q;+1/p-1/t11-,12-,13+,14+,15+,16+;/m0./s1

InChI key

DHARCCXVEUOMEO-JQQQFMHGSA-M

Unit Definition

One A265 unit = approx. 20 μg oligonucleotide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service