Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

O5005

Sigma-Aldrich

cis-6,9,12,15-Octadecatetraenoic acid sodium salt

~90% (GC)

Synonym(s):

Stearidonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C18H27O2Na
CAS Number:
Molecular Weight:
298.40
MDL number:
UNSPSC Code:
12352211

Assay

~90% (GC)

storage temp.

−20°C

SMILES string

[Na].CC\C=C\C\C=C\C\C=C\C\C=C\CCCCC(O)=O


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service