Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

O0875

Sigma-Aldrich

Butyl oleate

~75%

Synonym(s):

Oleic acid butyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H42O2
CAS Number:
Molecular Weight:
338.57
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

~75%

density

0.88 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCCCCCC\C=C/CCCCCCCC(=O)OCCCC

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan Mou et al.
Molecules (Basel, Switzerland), 20(11), 20320-20333 (2015-11-17)
The endophytic fungus Berkleasmium sp. Dzf12, isolated from Dioscorea zingiberensis, was found to produce palmarumycins C12 and C13 which possess a great variety of biological activities. Seven biocompatible water-immiscible organic solvents including n-dodecane, n-hexadecane, 1-hexadecene, liquid paraffin, dibutyl phthalate, butyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service