Skip to Content
MilliporeSigma
All Photos(1)

Documents

N9755

Sigma-Aldrich

Bis(4-nitrophenyl) carbonate

Synonym(s):

4-Nitrophenyl carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2NC6H4O)2CO
CAS Number:
Molecular Weight:
304.21
Beilstein:
1892897
EC Number:
MDL number:
UNSPSC Code:
12000000

mp

136-139 °C (lit.)

SMILES string

[O-][N+](=O)c1ccc(OC(=O)Oc2ccc(cc2)[N+]([O-])=O)cc1

InChI

1S/C13H8N2O7/c16-13(21-11-5-1-9(2-6-11)14(17)18)22-12-7-3-10(4-8-12)15(19)20/h1-8H

InChI key

ACBQROXDOHKANW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for the synthesis of 4-nitrophenyl active esters of amino acids. Used in the preparation of symmetrical and unsymmetrical ureas.

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service