Skip to Content
MilliporeSigma
All Photos(1)

Documents

N9642

Sigma-Aldrich

β-(2-Naphthyl)-D-Ala-Cys-Tyr-D-Trp-Lys-Val-Cys-Thr amide [Disulfide Bridge: 2–7]

≥97% (HPLC)

Synonym(s):

BIM-23014C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C54H69N11O10S2
CAS Number:
Molecular Weight:
1096.32
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CC(C)C1NC(=O)C(CCCCN)NC(=O)C(Cc2c[nH]c3ccccc23)NC(=O)C(Cc4ccc(O)cc4)NC(=O)C(CSSCC(NC1=O)C(=O)NC(C(C)O)C(N)=O)NC(=O)C(N)Cc5ccc6ccccc6c5

Gene Information

Biochem/physiol Actions

sst2 somatostatin receptor agonist.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Piwko et al.
Naunyn-Schmiedeberg's archives of pharmacology, 352(6), 607-613 (1995-12-01)
Radioligand binding studies were performed in membranes of rat cerebellum using [125I]-[Tyr3]octreotide ([125I]204-090) to characterize the nature of cerebellar somatostatin receptors. Saturation experiments suggest the presence of a single class of binding sites with high affinity, pKd = 9.53 +/-
J E Taylor et al.
Biochemical and biophysical research communications, 153(1), 81-86 (1988-05-31)
An endocrinologically-potent octapeptide analogue of somatostatin (SRIF), 3-(2-naphthyl)-D-Ala-Cys-Tyr-D-Trp-Lys-Val-Cys-Thr-NH2 (BIM-23014 C), was examined for its ability to inhibit the in vitro and in vivo growth of the human small cell lung carcinoma (SCLC) line, NCI-H69. When cultured cells were implanted into

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service