Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N9256

Sigma-Aldrich

2-Nitrophenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4OH
CAS Number:
Molecular Weight:
139.11
Beilstein:
775403
EC Number:
MDL number:
UNSPSC Code:
12352200

vapor pressure

1 mmHg ( 49.3 °C)

SMILES string

Oc1ccccc1[N+]([O-])=O

InChI

1S/C6H5NO3/c8-6-4-2-1-3-5(6)7(9)10/h1-4,8H

InChI key

IQUPABOKLQSFBK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

212.9 °F - closed cup

Flash Point(C)

100.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service