Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N8898

Sigma-Aldrich

Nerve Growth Factor Receptor/Fc Chimera human

>95% (SDS-PAGE), recombinant, expressed in baculovirus infected Sf21 cells, lyophilized powder

Synonym(s):

NGF R, p75 Neurotrophin receptor

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

Assay

>95% (SDS-PAGE)

form

lyophilized powder

potency

0.885 μg/mL ED50

mol wt

75 kDa

packaging

pkg of 50 μg

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... NGFR(4804)

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing 2.5 mg bovine serum albumin.

Analysis Note

The biological activity is measured by its ability to inhibit NGF-induced proliferation of TF-1 cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service