Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

N8883

Sigma-Aldrich

6-Nitro-3-(phenylacetamido)benzoic acid

≥98%

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$272.00
100 MG
$819.00

$272.00


Please contact Customer Service for Availability


Select a Size

Change View
25 MG
$272.00
100 MG
$819.00

About This Item

Empirical Formula (Hill Notation):
C15H12N2O5
CAS Number:
Molecular Weight:
300.27
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

$272.00


Please contact Customer Service for Availability

Quality Level

Assay

≥98%

form

powder

solubility

ethanol: 9.80-10.20 mg/mL, clear, faintly yellow to yellow

storage temp.

2-8°C

SMILES string

OC(=O)c1cc(NC(=O)Cc2ccccc2)ccc1[N+]([O-])=O

InChI

1S/C15H12N2O5/c18-14(8-10-4-2-1-3-5-10)16-11-6-7-13(17(21)22)12(9-11)15(19)20/h1-7,9H,8H2,(H,16,18)(H,19,20)

InChI key

QHVQEQRGDKOHHC-UHFFFAOYSA-N

General description

6-Nitro-3-(phenylacetamido)benzoic acid is a chromogenic analog of penicillin.[1]

Substrates

Substrate for penicillin amidase.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Davide A Cecchini et al.
BMC biotechnology, 7, 54-54 (2007-09-12)
Immobilized Penicillin G Acylase (PGA) derivatives are biocatalysts that are industrially used for the hydrolysis of Penicillin G by fermentation and for the kinetically controlled synthesis of semi-synthetic beta-lactam antibiotics. One of the most used supports for immobilization is glyoxyl-activated
Preparation and general properties of crystalline penicillin acylase from Escherichia coli ATCC 11 105.
C Kutzbach et al.
Hoppe-Seyler's Zeitschrift fur physiologische Chemie, 355(1), 45-53 (1974-01-01)
Q J Zhang et al.
Analytical biochemistry, 156(2), 413-416 (1986-08-01)
A simple, rapid assay for screening penicillin G acylase-producing bacteria is presented. The method is based on the formation of yellow 2-nitro-5-aminobenzoic acid by penicillin G acylase acting on 2-nitro-5-phenylacetaminobenzoic acid (NIPAB). NIPAB test paper is briefly applied to bacterial
Wynand B L Alkema et al.
European journal of biochemistry, 269(8), 2093-2100 (2002-05-03)
Penicillin acylase of Escherichia coli catalyses the hydrolysis and synthesis of beta-lactam antibiotics. To study the role of hydrophobic residues in these reactions, we have mutated three active-site phenylalanines. Mutation of alphaF146, betaF24 and betaF57 to Tyr, Trp, Ala or
Direct spectrophotometric measurement of enzyme activity in heterogeneous systems with insoluble substrate or immobilized enzyme.
B Galunsky et al.
Analytical biochemistry, 221(1), 213-214 (1994-08-15)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service