Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N8636

Sigma-Aldrich

Neocarratetraose 41-sulfate sodium salt

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C24H37O22SNa
CAS Number:
Molecular Weight:
732.59
MDL number:
UNSPSC Code:
12352201
Pricing and availability is not currently available.

Assay

≥98%

storage temp.

2-8°C

SMILES string

[Na+].OC[C@H]1O[C@@H](OC2[C@H]3CO[C@@H]2[C@@H](O)[C@H](O3)O[C@@H]4[C@@H](O)[C@H](O)O[C@H](CO)[C@@H]4OS([O-])(=O)=O)[C@H](O)[C@@H](O[C@H]5O[C@@H]6CO[C@@H](C6O)[C@H]5O)[C@H]1O

Other Notes

Mixed anomers

Preparation Note

Prepared enzymatically from κ-carrageenan.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M W McLean et al.
European journal of biochemistry, 101(2), 497-505 (1979-11-01)
A glycosulphatase present in the soluble fraction of disrupted Pseudomonas carrageenovora has been purified 500-fold by gel filtration on Sephacryl S-200 and ion-exchange chromatography on DEAE-Sepharose CL-6B. By dodecylsulphate/polyacrylamide gel electrophoresis the enzyme is practically homogeneous and has a molecular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service