Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

N7257

Sigma-Aldrich

Naphthol Green B

Technical grade

Synonym(s):

Acid Green 1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H15FeN3Na3O15S3
CAS Number:
Molecular Weight:
878.46
Colour Index Number:
10020
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

powder

λmax

714 nm

SMILES string

[Na+].[Na+].[Na+].[O-]S(=O)(=O)c1ccc2c(N=O)c(O[Fe](Oc3ccc4cc(ccc4c3N=O)S([O-])(=O)=O)Oc5ccc6cc(ccc6c5N=O)S([O-])(=O)=O)ccc2c1

InChI

1S/3C10H7NO5S.Fe.3Na/c3*12-9-4-1-6-5-7(17(14,15)16)2-3-8(6)10(9)11-13;;;;/h3*1-5,12H,(H,14,15,16);;;;/q;;;+3;3*+1/p-6

InChI key

JMXROTHPANUTOJ-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service