Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N6659

Sigma-Aldrich

NFκB Control Peptide

p65, human, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$280.00

$280.00


Please contact Customer Service for Availability


Select a Size

Change View
50 μG
$280.00

About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

$280.00


Please contact Customer Service for Availability

Product Name

NFκB (p65) Control Peptide human, ~1 mg/mL, buffered aqueous solution

biological source

human

Quality Level

form

buffered aqueous solution

concentration

~1 mg/mL

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... RELA(5970)

Amino Acid Sequence

Met-Asn-Glu-Leu-Phe -Pro-Leu-Ile-Phe-Pro-Ala-Glu-Pro-Ala-Glu-Ala-Ser-Gly-Pro-Tyr-Val

Application

For use as a blocking peptide for anti-NFκB(p65) subunit.

Physical form

Solution containing 0.01% sodium azide.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service