Skip to Content
Merck
All Photos(1)

Key Documents

N6129

Sigma-Aldrich

3-Nitrophenyl α-D-galacto­pyran­oside

≥98%

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$611.25
1 G
$1,222.50
2 G
$2,261.25

Vendor SKU: 3D-033-500MG

$611.25


Ships in 21 days - from Aldrich Partner.


Select a Size

Change View
500 MG
$611.25
1 G
$1,222.50
2 G
$2,261.25

About This Item

Empirical Formula (Hill Notation):
C12H15NO8
CAS Number:
Molecular Weight:
301.25
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

Vendor SKU: 3D-033-500MG

$611.25


Ships in 21 days - from Aldrich Partner.

Assay

≥98%

storage temp.

−20°C

SMILES string

OCC1OC(Oc2cccc(c2)N(=O)=O)C(O)C(O)C1O

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Don't see the Right Version?

    If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service