Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N4912

Sigma-Aldrich

NMA-Gly-Cys-Gly-Val-Leu-Leu-Lys(dnp)-Arg-Arg-NH2 trifluoroacetate salt

≥95% (HPLC), solid

Synonym(s):

NMA-GCGVLLK(dnp)RR-NH2

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$364.00

$364.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$364.00

About This Item

Empirical Formula (Hill Notation):
C56H90N20O14S · xC2HF3O2
Molecular Weight:
1299.50 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$364.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥95% (HPLC)

form

solid

storage condition

desiccated
protect from light
under inert gas

color

yellow

solubility

H2O: ≥0.5 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

FC(F)(F)C(=O)O.SC[C@H](NC(=O)CNC(=O)[C@@H](NC)CC(=O)O)C(=O)NCC(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCCCNc1c(cc(cc1)[N+](=O)[O-])[N+](=O)[O-])C(=O)N[C@@H](CCCNC(=N)N)C(=O)N[C@@H](CCCNC(=N)N)C(=O)N

InChI key

RHLODBFQRYLALJ-ZWIFCGJVSA-N

Amino Acid Sequence

Nme-Asp-Gly-Cys-Gly-Val-Leu-Leu-Lys-DNP-Arg-Arg-NH2

Biochem/physiol Actions

Notch 1 Peptide, fluorogenic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshihito Taniguchi et al.
Proceedings of the National Academy of Sciences of the United States of America, 99(6), 4014-4019 (2002-03-14)
Notch receptors undergo three distinct proteolytic cleavages during maturation and activation. The third cleavage occurs within the plasma membrane and results in the release and translocation of the intracellular domain into the nucleus to execute Notch signaling. This so-called gamma-secretase

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service