Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N4413

Sigma-Aldrich

Anti-Nanog antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-FLJ12581, Anti-Nanog homeobox

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

concentration

~1 mg/mL

technique(s)

western blot: 1:1,000-1:2,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NANOG(79923)
mouse ... Nanog(71950)
rat ... Nanog(414065)

General description

Nanog homeobox (NANOG) is a 305 amino acid transcription factor. It is expressed in many types of cancers.

Immunogen

synthetic peptide corresponding to an internal sequence of human Nanog.

Biochem/physiol Actions

Nanog homeobox (NANOG) works with pluripotency factors like sex determining region Y-box 2 (SOX2). It is involved in conserving the pluripotency and self-renewal properties of embryonic stem cells. NANOG also associates with the c-Jun promoter.

Physical form

solution in phosphate buffered saline, containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

NANOG upregulates c-Jun oncogene expression through binding the c-Jun promoter.
Lin Y
Molecular Carcinogenesis, 54(11), 1407-1416 (2015)
Shuai Gong et al.
Molecular carcinogenesis, 54(9), 679-687 (2015-05-28)
As one of the key pluripotency transcription factors, NANOG plays a critical role in maintaining the self-renewal and pluripotency in normal embryonic stem cells. Recent data indicate that NANOG is expressed in a variety of cancers and its expression correlates
Structure-based discovery of NANOG variant with enhanced properties to promote self-renewal and reprogramming of pluripotent stem cells.
Hayashi Y
Proceedings of the National Academy of Sciences of the USA, 112(15), 4666-4671 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service