Skip to Content
MilliporeSigma
All Photos(1)

Documents

N4255

Sigma-Aldrich

α-Nicotinamide mononucleotide

≥98%

Synonym(s):

α-NMN, α-Nicotinamide ribose monophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15N2O8P
CAS Number:
Molecular Weight:
334.22
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

Assay

≥98%

storage temp.

−20°C

SMILES string

NC(=O)C1=CC=C[N](=C1)C2OC(COP(O)(O)=O)C(O)C2O

Caution

This is not the common NMN required in the usual enzyme systems.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service