Skip to Content
MilliporeSigma
All Photos(1)

Documents

N3759

Sigma-Aldrich

3-Nitro-1H-1,2,4-triazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H2N4O2
CAS Number:
Molecular Weight:
114.06
Beilstein:
607167
EC Number:
MDL number:
UNSPSC Code:
12352103

mp

210 °C (dec.) (lit.)

SMILES string

[O-][N+](=O)c1nc[nH]n1

Looking for similar products? Visit Product Comparison Guide

Packaging

Packaged under Argon and sealed with PTFE-faced septum.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service