Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N2509

Sigma-Aldrich

2-Nitrophenyl 1-thio-β-D-galactopyranoside

for detection of lac mutants

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15NO7S
CAS Number:
Molecular Weight:
317.32
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

grade

for molecular biology

form

powder

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](Sc2ccccc2[N+]([O-])=O)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C12H15NO7S/c14-5-7-9(15)10(16)11(17)12(20-7)21-8-4-2-1-3-6(8)13(18)19/h1-4,7,9-12,14-17H,5H2/t7-,9+,10+,11-,12+/m1/s1

InChI key

SZAOZNVCHHBUDZ-RUXWNWLUSA-N

General description

2-Nitrophenyl 1-thio-β-D-galactopyranoside is an enzyme substrate used in the detection of lac- mutants.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D S Gregerson et al.
Journal of immunology (Baltimore, Md. : 1950), 163(2), 1073-1080 (1999-07-08)
Recent studies revealing active mechanisms of immune privilege in neural tissues have diminished the putative role of passive tolerance. To examine the significance of Ag localization in the retina on immune privilege, the immune responses of transgenic mice expressing high
Ausubel, F.M., et al.
Current Protocols in Molecular Biology, 1-1 (1998)
S Gharbi et al.
Biochimie, 68(10-11), 1159-1163 (1986-10-01)
Mutants altered in their expression of the hybrid pepN-lacZ gene have been selected for resistance to p-nitrophenyl-beta-D-thiogalactopyranoside (a bacteriostatic compound that enters the cells via lac permease). A unique mutation decreasing the level of pepN expression to 9% of that
M T Martins et al.
Applied and environmental microbiology, 58(9), 3095-3100 (1992-09-01)
A total of 449 Escherichia coli isolates in treated and raw water sources were submitted to DNA-DNA hybridization using seven different DNA probes to detect homology to sequences that code for Shiga-like toxins I and II; heat-stabile and heat-labile toxins

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service