Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N218

Sigma-Aldrich

Monoclonal Anti-Nitric Oxide Synthase, Universal antibody produced in mouse

clone NOS-3F7-B11-B5, ascites fluid

Synonym(s):

Anti-IHPS1, Anti-N-NOS, Anti-NC-NOS, Anti-NOS, Anti-bNOS, Anti-nNOS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

NOS-3F7-B11-B5, monoclonal

species reactivity

mouse, bovine, rat

technique(s)

immunohistochemistry (frozen sections): 1:100
western blot: 1:50-1:500

isotype

IgM

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Nos1(18125)
rat ... Nos1(24598)

Specificity

The antibody reacts with iNOS, eNOS and nNOS from rat and bovine, and with iNOS in induced RAW 264.7 mouse macrophages. In immunoblotting, the antibody detects a band at 130 kDa representing iNOS in samples first induced with IFN (interferon) and LPS (lipopolysaccharides). It also detects a band at 155 kDa in tissues expressing nNOS and a band at 140 kDa in tissues expressing eNOS. The antibody may be used to localize and detect the three isoforms of nitric oxide synthase (NOS): neuronal or brain (nNOS or bNOS), endothelial (eNOS) and inducible (iNOS).

Immunogen

bovine cerebellum NOS.

Biochem/physiol Actions

Neuronal nitric acid synthase (Nos1) in involved in the production of nitric oxide. Mutations in the gene encoding it increases the susceptibility to behavioral dysfunction after an exposure to alcohol in mice.

Physical form

Ascites fluid containing ≤0.1% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bahri Karacay et al.
Alcoholism, clinical and experimental research, 39(2), 212-220 (2015-02-17)
Alcohol abuse during pregnancy often induces neuropsychological problems in the offspring, including learning disorders, attention deficits, and behavior problems, all of which are prominent components of fetal alcohol spectrum disorders (FASD). However, not all children who were exposed to alcohol
Krystyna Makowska et al.
International journal of molecular sciences, 22(19) (2021-10-14)
Bisphenol A (BPA) is a substance used in the manufacture of plastics which shows multidirectional adverse effects on living organisms. Since the main path of intoxication with BPA is via the gastrointestinal (GI) tract, the stomach and intestine are especially
Liliana Rytel
International journal of molecular sciences, 19(10) (2018-10-03)
Bisphenol A (BPA), a substance commonly used in the manufacture of plastics, shows multidirectional negative effects on humans and animals. Due to similarities to estrogens, BPA initially leads to disorders in the reproductive system. On the other hand, it is
Bahri Karacay et al.
Alcoholism, clinical and experimental research, 39(2), 221-231 (2015-02-11)
Prenatal alcohol exposure can kill developing neurons, leading to microencephaly and mental retardation. However, not all fetuses are equally vulnerable to alcohol's neurotoxic effects. While some fetuses are severely affected and are ultimately diagnosed with fetal alcohol syndrome (FAS), others
Liliana Rytel et al.
International journal of molecular sciences, 21(12) (2020-07-02)
Bisphenol A (BPA) is one of the most common environmental pollutants among endocrine disruptors. Due to its similarity to estrogen, BPA may affect estrogen receptors and show adverse effects on many internal organs. The reproductive system is particularly vulnerable to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service