Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

N1626

Sigma-Aldrich

Neocuproine hydrochloride hydrate

BioReagent, crystalline

Synonym(s):

2,9-Dimethyl-1,10-phenanthroline hydrochloride hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$245.00
25 G
$788.00

$245.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$245.00
25 G
$788.00

About This Item

Empirical Formula (Hill Notation):
C14H12N2 · HCl · xH2O
CAS Number:
Molecular Weight:
244.72 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

$245.00


Please contact Customer Service for Availability

Request a Bulk Order

product line

BioReagent

form

crystalline

mp

250 °C (dec.) (lit.)

suitability

suitable for determination of copper

SMILES string

O.Cl.Cc1ccc2ccc3ccc(C)nc3c2n1

InChI

1S/C14H12N2.ClH.H2O/c1-9-3-5-11-7-8-12-6-4-10(2)16-14(12)13(11)15-9;;/h3-8H,1-2H3;1H;1H2

InChI key

APDYLFLZNGECIM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tatjana Kochanė et al.
International journal of biological macromolecules, 144, 544-552 (2019-12-22)
A series of modified poly(urethane-urea) carriers for immobilization of maltogenase from Bacillus stearothermophilus were presented. Poly(urethane-urea) microparticles (PUU) from poly(vinyl alcohol) and isophorone diisocyanate were modified with ethylenediamine, glutaraldehyde, trichlorotriazine and gold nanoparticles (AuNPs). Covalent attachment of enzyme to carrier
Jong-Min Park et al.
Biosensors & bioelectronics, 71, 306-312 (2015-04-29)
This work presented a highly sensitive bacterial antibiotic susceptibility test through β-lactamase assay using Parylene-matrix chip. β-lactamases (EC 3.5.2.6) are an important family of enzymes that confer resistance to β-lactam antibiotics by catalyzing the hydrolysis of these antibiotics. Here we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service