Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N1145

Sigma-Aldrich

1-Naphthaleneacetic acid potassium salt

BioReagent, suitable for plant cell culture, ≥95%

Synonym(s):

α-Naphthaleneacetic acid, NAA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C12H9O2K
CAS Number:
Molecular Weight:
224.30
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

product line

BioReagent

Assay

≥95%

technique(s)

cell culture | plant: suitable

application(s)

agriculture

SMILES string

[K].OC(=O)Cc1cccc2ccccc12

InChI

1S/C12H10O2.K.H/c13-12(14)8-10-6-3-5-9-4-1-2-7-11(9)10;;/h1-7H,8H2,(H,13,14);;

InChI key

VDSVVXDRPHZMEX-UHFFFAOYSA-N

Gene Information

Application

1-Naphthaleneacetic acid (NAA) is a synthetic phytohormone auxin that is added to cell culture media such as Murashige & Skoog media and Chu′s N6 media.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evelyn E Schwager et al.
BMC biology, 15(1), 62-62 (2017-08-02)
The duplication of genes can occur through various mechanisms and is thought to make a major contribution to the evolutionary diversification of organisms. There is increasing evidence for a large-scale duplication of genes in some chelicerate lineages including two rounds
Angela Maria Amorini et al.
Journal of cellular and molecular medicine, 21(3), 530-542 (2016-10-04)
In this study, concentrations of free amino acids (FAA) and amino group containing compounds (AGCC) following graded diffuse traumatic brain injury (mild TBI, mTBI; severe TBI, sTBI) were evaluated. After 6, 12, 24, 48 and 120 hr aspartate (Asp), glutamate
O Rahul Patharkar et al.
Plant physiology, 172(1), 510-520 (2016-07-30)
Drought-triggered abscission is a strategy used by plants to avoid the full consequences of drought; however, it is poorly understood at the molecular genetic level. Here, we show that Arabidopsis (Arabidopsis thaliana) can be used to elucidate the pathway controlling
Hisayo Yamane et al.
PloS one, 14(4), e0214788-e0214788 (2019-04-10)
Most deciduous fruit trees cultivated in the temperate zone require a genotype-dependent amounts of chilling exposure for dormancy release and bud break. In Japanese apricot (Prunus mume), DORMANCY-ASSOCIATED MADS-box 6 (PmDAM6) may influence chilling-mediated dormancy release and bud break. In
Gregg W C Thomas et al.
Genome biology, 21(1), 15-15 (2020-01-24)
Arthropods comprise the largest and most diverse phylum on Earth and play vital roles in nearly every ecosystem. Their diversity stems in part from variations on a conserved body plan, resulting from and recorded in adaptive changes in the genome.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service