Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N1140

SAFC

NCTC 109 Medium

With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture

Pharma Manufacturing

Synonym(s):

109 Cell Culture Medium, NCTC Medium

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
$74.90
6 X 500 ML
$467.00

$74.90


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 ML
$74.90
6 X 500 ML
$467.00

About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

$74.90


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

glucose: 1.0 g/L
phenol red: 0.02 g/L
L-glutamine: no
NaHCO3: 2.2 g/L

shipped in

ambient

storage temp.

2-8°C

General description

NCTC-109 and the 135 modification were formulated to establish and maintain a strain of mouse cells (L929) in a chemically defined and serum free medium.

Reconstitution

Supplement with 0.14 g/L L-glutamine.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

NCTC 109 Medium Formulation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service