MTCR1070
[MTC"] Bio Tube Rack
to hold, 30 x 15 mL and 20 x 50 mL tubes, orange, pack of 5 ea
Synonym(s):
laboratory rack, stacking western blot box, tube holder
All Photos(1)
About This Item
Recommended Products
material
orange
polypropylene
feature
to hold 30 x 15 mL and 20 x 50 mL tubes
packaging
pack of 5 ea
manufacturer/tradename
MTC™ Bio R1070
Related Categories
General description
Rack for 30x15 & 20x50ml (17x20x6cm), Orange, 5/pk
Features and Benefits
- Polypropylene, withstand -80°C
- Compact and space efficient
- Autoclavable
Legal Information
MTC is a trademark of MTC Bio