Skip to Content
MilliporeSigma
All Photos(1)

Documents

MTCP7005

MTC Bio ASPIR-8 Resevoir

capacity 5 mL, sterile, pack of 300 ea (bulk bag)

Synonym(s):

plastic tube, reagent dish, reagent holder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121522
NACRES:
NB.15

sterility

sterile

packaging

pack of 300 ea (bulk bag)

manufacturer/tradename

MTC Bio P7005

capacity

5 mL

Looking for similar products? Visit Product Comparison Guide

General description

ASPIR-8 Reservoir, 5mL, non-sterile, bulk pack, 300/cs

Features and Benefits

  • Graduated inner walls
  • Micro-trough U-shaped bottoms
  • Pouring spouts on all 4 corners

Legal Information

MTC is a trademark of MTC Bio

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service